EMAIL THIS PAGE TO A FRIEND
ACS Applied Materials & Interfaces
Highly sensitive reduced graphene oxide impedance sensor harnessing ?-stacking interaction mediated direct deposition of protein probes.
PMID 23551147
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
302031
Trifluoroacetic acid, for HPLC, ≥99.0%
C2HF3O2