EMAIL THIS PAGE TO A FRIEND
The Consultant pharmacist : the journal of the American Society of Consultant Pharmacists
Antacid-induced acute pancreatitis.
PMID 23552706
Abstract
To report a case of acute pancreatitis associated with the administration of high doses of over-the-counter (OTC) antacids containing calcium carbonate. An acute care 276-bed community hospital. In this case report, a 68-year-old male was admitted to the hospital for nausea, vomiting, and stomach pain. Patient reported taking 15-20 OTC calcium carbonate (4.5-6.0 g elemental calcium) tablets for four to five days prior to admission. Patient's calcium level upon admission was 18.5 mg/dL. Laboratory values of amylase and lipase peaked at 2,768 U/L and 7,091 U/L, respectively. An abdominal computed tomography confirmed acute pancreatitis. For a period of 45 days, treatment in the intensive care unit for acute pancreatitis and latter for necrotizing pancreatitis was conducted. This case study supports an association between hypercalcemia and pancreatitis in patients who take greater than recommended dosages of calcium-containing antacids. OTC antacids are popular, but can cause serious adverse events when not used as directed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6763
Calcium carbonate, ReagentPlus®
CCaO3
239216
Calcium carbonate, ACS reagent, ≥99.0%, powder
CCaO3
12010
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
CCaO3
900500
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
CCaO3
62287 Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
L4777
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
62288 Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
62301 Lipase from Aspergillus niger, powder (fine), ~200 U/g
62285 Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
75577 Lipase from Burkholderia sp., lyophilized powder, slightly beige, ~12 U/mg
02569 Lipase from Candida antarctica, lyophilized, powder, beige, ~0.3 U/mg
62302 Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
L1754 Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
62316 Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
L8525 Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
L3170
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
L8906 Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
L9031 Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
62298 Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
L0382 Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
62309 Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
28602 Lipase from Pseudomonas fluorescens, powder, slightly beige, ≥160 U/mg
L9518
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
L4277 Lipase from Rhizomucor miehei, ≥20,000 U/g
62310 Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
62305 Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
L0777 Lipase from Thermomyces lanuginosus, solution, ≥100,000 U/g
L3001 Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg protein
62306 Lipase from wheat germ, ~0.1 U/mg
73940 Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg