EMAIL THIS PAGE TO A FRIEND
Journal of the Science of Food and Agriculture
Combined effect of glycation and sodium carbonate-bicarbonate buffer concentration on IgG binding, IgE binding and conformation of ovalbumin.
PMID 23553593