EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Single-molecule detection of 5-hydroxymethylcytosine in DNA through chemical modification and nanopore analysis.
PMID 23559386