EMAIL THIS PAGE TO A FRIEND
Liver international : official journal of the International Association for the Study of the Liver
Status of research on vitamin D supplementation in treating or preventing liver fibrosis.
PMID 23560728