EMAIL THIS PAGE TO A FRIEND
Gene
Wacław Szybalski's contribution to immunotherapy: HGPRT mutation & HAT selection as first steps to gene therapy and hybrid techniques in mammalian cells.
PMID 23562724
Abstract
In this report we describe Wacław Szybalski's fundamental contribution to gene therapy and immunotherapy. His 1962 PNAS paper (Szybalska and Szybalski, 1962) documented the first successful gene repair in mammalian cells. Furthermore, this was also the first report on the HAT selection method used later in many applications. Most importantly, somatic cell fusion and HAT selection were subsequently used to develop monoclonal antibody technology, which contributed significantly to the progress of today's medicine.