EMAIL THIS PAGE TO A FRIEND
Nigerian journal of clinical practice
Paraphenylenediamine poisoning.
PMID 23563473
Abstract
Hair dye containing paraphenylenediamine (PPD) is widely used in India because of its free availability and low cost. PPD produces local as well as systemic toxic effects when applied topically and/or ingested. It is highly toxic when taken by mouth and the outcome depends mainly on the dose taken. Important clinical manifestations are angioedema leading to dysphasia and respiratory distress, rhabdomyolysis, intravascular hemolysis, acute renal failure and hepatic necrosis. Myocarditis or fatal arrhythmia may also occur in PPD poisoning. Mainstay of management is early recognition and supportive measures as there is no specific antidote. We hereby report a young female who presented to us with features of angioedema, cardiac manifestation and hepatic dysfunction after ingesting PPD, which was treated successfully. In the absence of laboratory facilities, clinical features like angioedema and chocolate brown-colored urine could be suggestive of PPD poisoning.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78460
1,4-Phenylenediamine dihydrochloride, ≥99.0%
C6H8N2 · 2HCl