EMAIL THIS PAGE TO A FRIEND
Nature chemical biology
A single-molecule analysis reveals morphological targets for cellulase synergy.
PMID 23563526
Abstract
The mechanisms of enzyme activity on solid substrates are not well understood. Unlike enzyme catalysis in aqueous solutions, enzyme activity on surfaces is complicated by adsorption steps and structural heterogeneities that make enzyme-substrate interactions difficult to characterize. Cellulase enzymes, which catalyze the depolymerization of cellulose, show binding specificities for different cellulose surface morphologies, but the influence of these specificities on the activity of multienzyme mixtures has remained unclear. We developed a metric to quantify binding-target arrangements determined by photoactivated localization microscopy, and we used that metric to show that combinations of cellulases designed to bind within similar but nonidentical morphologies can have synergistic activity. This phenomenon cannot be explained with the binary crystalline or amorphous classifications commonly used to characterize cellulase-binding targets. Our results reveal a strategy for improving the activity of cellulolytic mixtures and demonstrate a versatile method for investigating protein organization on heterogeneous surfaces.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6429
α-Cellulose, BioReagent, suitable for insect cell culture
11363
Avicel® PH-101, tested according to Ph.Eur.
11365
Avicel® PH-101, ~50 μm particle size
C1184 Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
22178 Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
C2605
Cellulase from Aspergillus sp., aqueous solution
C2730 Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
C8546
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
C0615
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid
C1794 Cellulase from Trichoderma sp., BioReagent, plant cell culture tested, 3-10 units/mg solid
C9499
Cellulase, thermostable from Clostridium thermocellum, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
22182
Cellulose, acid washed, from spruce, for column chromatography
22184
Cellulose, acid washed, powder, for column chromatography
22183
Cellulose, powder, for column chromatography
22197
Cellulose, DS-0, powder, for thin layer chromatography
09906
Cellulose, DFS-0, microcrystalline, for thin layer chromatography
435244
Cellulose, colloidal, microcrystalline
310697
Cellulose, microcrystalline, powder, 20 μm
435236
Cellulose, microcrystalline, powder
S6790
Sigmacell Cellulose, Type 101, Highly purified, fibers
C6413
Cellulose, microgranular