EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica Sinica
Functional comparison of the reverse mode of Na+/Ca2+ exchangers NCX1.1 and NCX1.5 expressed in CHO cells.
PMID 23564083
Abstract
To investigate the reverse mode function of Na(+)/Ca(2+) exchangers NCX1.1 and NCX1.5 expressed in CHO cells as well as their modulations by PKC and PKA. CHO-K1 cells were transfected with pcDNA3.1 (+) plasmid carrying cDNA of rat cardiac NCX1.1 and brain NCX1.5. The expression of NCX1.1 and NCX1.5 was examined using Western blot analysis. The intracellular Ca(2+) level ([Ca(2+)]i) was measured using Ca(2+) imaging. Whole-cell NCX currents were recorded using patch-clamp technique. Reverse mode NCX activity was elicited by perfusion with Na(+)-free medium. Ca(2+) paradox was induced by Ca(2+)-free EBSS medium, followed by Ca(2+)-containing solution (1.8 or 3.8 mmol/L CaCl2). The protein levels of NCX1.1 and NCX1.5 expressed in CHO cells had no significant difference. The reverse modes of NCX1.1 and NCX1.5 in CHO cells exhibited a transient increase of [Ca(2+)]i, which was followed by a Ca(2+) level plateau at higher external Ca(2+) concentrations. In contrast, the wild type CHO cells showed a steady increase of [Ca(2+)]i at higher external Ca(2+) concentrations. The PKC activator PMA (0.3-10 μmol/L) and PKA activator 8-Br-cAMP (10-100 μmol/L) significantly enhanced the reverse mode activity of NCX1.1 and NCX1.5 in CHO cells. NCX1.1 was 2.4-fold more sensitive to PKC activation than NCX1.5, whereas the sensitivity of the two NCX isoforms to PKA activation had no difference. Both PKC- and PKA-enhanced NCX reverse mode activities in CHO cells were suppressed by NCX inhibitor KB-R7943 (30 μmol/L). Both NCX1.1 and NCX1.5 are functional in regulating and maintaining stable [Ca(2+)]i in CHO cells and differentially regulated by PKA and PKC. The two NCX isoforms might be useful drug targets for heart and brain protection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K4144
KB-R7943, ≥98% (HPLC), powder
C16H17N3O3S·CH3SO3H