EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
CD44 targeting magnetic glyconanoparticles for atherosclerotic plaque imaging.
PMID 23568520
Abstract
The cell surface adhesion molecule CD44 plays important roles in the initiation and development of atherosclerotic plaques. We aim to develop nanoparticles that can selectively target CD44 for the non-invasive detection of atherosclerotic plaques by magnetic resonance imaging. Magnetic glyconanoparticles with hyaluronan immobilized on the surface have been prepared. The binding of these nanoparticles with CD44 was evaluated in vitro by enzyme linked immunosorbent assay, flow cytometry and confocal microscopy. In vivo magnetic resonance imaging of plaques was performed on an atherosclerotic rabbit model. The magnetic glyconanoparticles can selectively bind CD44. In T2* weighted magnetic resonance images acquired in vivo, significant contrast changes in aorta walls were observed with a very low dose of the magnetic nanoparticles, allowing the detection of atherosclerotic plaques. Furthermore, imaging could be performed without significant delay after probe administration. The selectivity of hyaluronan nanoparticles in plaque imaging was established by several control experiments. Magnetic nanoparticles bearing surface hyaluronan enabled the imaging of atherosclerotic plaques in vivo by magnetic resonance imaging. The low dose of nanoparticles required, the possibility to image without much delay and the high biocompatibility are the advantages of these nanoparticles as contrast agents for plaque imaging.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3773
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
B5283
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
R2655
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
G2904
Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
A8062
Monoclonal Anti-Goat/Sheep IgG–Alkaline Phosphatase antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous glycerol solution
B3148
Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
F4891
Monoclonal Anti-Goat/Sheep IgG−FITC antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
A9452
Monoclonal Anti-Goat/Sheep IgG–Peroxidase antibody produced in mouse, clone GT-34, purified immunoglobulin, lyophilized powder
I6135
Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
I6260
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
I6760
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
F5016
Monoclonal Anti-Human IgG (Fc specific)−FITC antibody produced in mouse, clone HP-6017, purified immunoglobulin, buffered aqueous solution
I5885
Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
I9513
Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
I9888
Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
R1008
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
A2556
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
R3155
Monoclonal Anti-Rabbit IgG, Native−Peroxidase antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
R0761
Monoclonal Anti-Rat IgG2a antibody produced in mouse, clone R2A-2, ascites fluid
N1408 NGF-β human, from human, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture