EMAIL THIS PAGE TO A FRIEND
Clinical orthopaedics and related research
CORR Insights®: Surgical technique: Results of stabilization of sternoclavicular joint luxations using a polydioxanone envelope plasty.
PMID 23568678
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
719846
Resomer® X 206 S, poly(dioxanone)