EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Multifunctional three-dimensional macroporous nanoelectronic networks for smart materials.
PMID 23569270
Abstract
Seamless and minimally invasive integration of 3D electronic circuitry within host materials could enable the development of materials systems that are self-monitoring and allow for communication with external environments. Here, we report a general strategy for preparing ordered 3D interconnected and addressable macroporous nanoelectronic networks from ordered 2D nanowire nanoelectronic precursors, which are fabricated by conventional lithography. The 3D networks have porosities larger than 99%, contain approximately hundreds of addressable nanowire devices, and have feature sizes from the 10-μm scale (for electrical and structural interconnections) to the 10-nm scale (for device elements). The macroporous nanoelectronic networks were merged with organic gels and polymers to form hybrid materials in which the basic physical and chemical properties of the host were not substantially altered, and electrical measurements further showed a >90% yield of active devices in the hybrid materials. The positions of the nanowire devices were located within 3D hybrid materials with ∼14-nm resolution through simultaneous nanowire device photocurrent/confocal microscopy imaging measurements. In addition, we explored functional properties of these hybrid materials, including (i) mapping time-dependent pH changes throughout a nanowire network/agarose gel sample during external solution pH changes, and (ii) characterizing the strain field in a hybrid nanoelectronic elastomer structures subject to uniaxial and bending forces. The seamless incorporation of active nanoelectronic networks within 3D materials reveals a powerful approach to smart materials in which the capabilities of multifunctional nanoelectronics allow for active monitoring and control of host systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9539
Agarose, BioReagent, for molecular biology, low EEO
A6013
Agarose, Type I, low EEO
A6877
Agarose, Medium EEO, for molecular biology
A0169
Agarose, Type I-A, low EEO
A4718
Agarose, High resolution, for molecular biology
A4679
Agarose, Low EEO, for Immunoelectrophoresis
A7431
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
A9918
Agarose, Type II-A, Medium EEO
A0576
Agarose, Low EEO
A2790
Agarose, Wide range, for molecular biology
A9311
Agarose, Medium EEO, for electrophoresis
A2929
Agarose, For pulsed field electrophoresis running gel
A5030
Agarose, Ultra-low Gelling Temperature
A3643
Agarose, Type IV, Special High EEO
A2576
Agarose, Ultra-low Gelling Temperature, molecular biology grade
A9793
Agarose, High EEO
A7174
Agarose, High Gelling Temperature
A5304
Agarose, Special High EEO
A3054
Agarose, For pulsed field electrophoresis sample preparation
05066
Agarose, High EEO, for molecular biology
4B200
Sepharose® 4B, 45-165 μm bead diameter
6B100
Sepharose® 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
U0507
Ultrogel® A4, aqueous ethanol suspension, 60-140 μm (wet bead diameter), exclusion limit 20,000,000 Da, fractionation range 55,000-9,000,000 Da (globular proteins)