EMAIL THIS PAGE TO A FRIEND
International journal of chronic obstructive pulmonary disease
Inspiratory drive is related to dynamic pulmonary hyperinflation in COPD patients.
PMID 23569371
Abstract
Baseline high neuromuscular drive is present in chronic obstructive pulmonary disease (COPD). In moderate-to-very severe COPD patients, both static and/or dynamic pulmonary hyperinflation have been demonstrated at rest. To assess the influence of dynamic hyperinflation on neuromuscular drive at rest. We recruited 22 patients with severe-to-very severe COPD showing resting dynamic pulmonary hyperinflation, as assessed by the baseline reduction of inspiratory capacity (IC) (<80% of predicted). IC, occlusion pressure (P₀.₁), maximal inspiratory pressure (MIP), and their ratio were measured at end-expiratory lung volume (EELV) before and after acute inhalation of 400 mcg of albuterol (metered-dose inhaler plus spacer). In these patients the bronchodilator response was assessed also as lung volume changes. Only in COPD patients with a marked increase in IC (>12% of baseline and at least 200 mL) after bronchodilator, resting P0.1 showed a clinically significant decrease, despite the EELV diminution (P < 0.001). MIP was augmented following EELV reduction and therefore the P₀.₁/MIP ratio was markedly decreased (P < 0.001). In contrast, no changes in these indices were found after bronchodilator in COPD patients with insignificant variations of IC. Breathing pattern parameters did not vary in both sub-groups after albuterol. Following bronchodilator, significant P₀.₁ decrease, MIP increase, and reduction of the P₀.₁/MIP ratio were found only in COPD patients with a marked IC increase and these changes were closely related. These findings suggest that bronchodilators, by decreasing dynamic hyperinflation, may control exertional and/or chronic dyspnea partly through a reduction of central neuromuscular drive.