EMAIL THIS PAGE TO A FRIEND
Optics express
Slow light based on stimulated Raman scattering in an integrated liquid-core optical fiber filled with CS2.
PMID 23571972
Abstract
We demonstrate a fiber-based slow light system using a carbon disulfide (CS2)) filled integrated liquid-core optical fiber (i-LCOF). Using 1 meter of i-LCOF we were able to delay 18ps pulses up to 34ps; a delay of 188% of the pulse width. This experimental setup serves as a foundation for slow-light experiments in other nonlinear liquids. Numerical simulations of pulse-propagation equations confirmed the observed delay and a simplified method is presented that can be applied to calculate induced delay for non-cw Stokes pulses. The system is all-fiber and compact with delays greater than a pulse width, indicating potential application as an ultrafast controllable delay line for time division multiplexing in multiGb/s telecommunication systems.