EMAIL THIS PAGE TO A FRIEND
Mycoses
Misdiagnosed zoophile tinea faciei and tinea corporis effectively treated with isoconazole nitrate and diflucortolone valerate combination therapy.
PMID 23574022
Abstract
There have been few published reports on the human transmission of Trichophyton mentagrophytes, a zoophilic fungus frequently occurring in pets. Here we report on 2 girls, living with a pet dwarf rabbit, who presented with inflammatory skin lesions positive for T. mentagrophytes and subsequently diagnosed as zoophile tinea faciei and tinea corporis. The patients were successfully treated with systemic terbinafine and 2-week therapy with Travocort cream containing isoconazole nitrate 1% and diflucortolone valerate 0.1%.