EMAIL THIS PAGE TO A FRIEND
The Journal of chemical physics
Pre-selective screening for matrix elements in linear-scaling exact exchange calculations.
PMID 23574215
Abstract
We present a simple but accurate preselection method based on Schwarz integral estimates to determine the significant elements of the exact exchange matrix before its evaluation, thus providing an asymptotical linear-scaling behavior for non-metallic systems. Our screening procedure proves to be highly suitable for exchange matrix calculations on massively parallel computing architectures, such as graphical processing units, for which we present a first linear-scaling exchange matrix evaluation algorithm.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0512
Amylose from potato, used as amylase substrate