EMAIL THIS PAGE TO A FRIEND
British journal of industrial medicine
Studies of the carcinogenesis and tumorigenesis of skin applications of dodecylbenzene on hairless mice.
PMID 2357457
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
113239
1-Phenyldodecane, 97%
C18H30
44178
1-Phenyldodecane, analytical standard
C18H30