EMAIL THIS PAGE TO A FRIEND
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology
Can azathioprine 6-mercaptopurine treatment be withdrawn in patients with response in ulcerative colitis: what is the most appropriate time for this?
PMID 23576085