EMAIL THIS PAGE TO A FRIEND
Oxidative medicine and cellular longevity
Influence of block of NF-kappa B signaling pathway on oxidative stress in the liver homogenates.
PMID 23577221
Abstract
The aim of the present study was to assess whether BAY 11-7082, a nuclear factor-kappaB (NF- κ B) inhibitor, influences the level of reactive oxygen species (ROS), tumor necrosis factor alpha (TNF- α), and NF- κ B related signaling pathways in the liver. The animals were divided into 4 groups: I: saline; II: saline + endothelin-1 (ET-1) (1.25 μg/kg b.w., i.v.); III: saline + ET-1 (12.5 μg/kg b.w., i.v.); and IV: BAY 11-7082 (10 mg/kg b.w., i.v.) + ET-1 (12.5 μg/kg b.w., i.v.). Injection of ET-1 alone at a dose of 12.5 μg/kg b.w. showed a significant (P < 0.001) increase in thiobarbituric acid reactive substances (TBARS) and hydrogen peroxide (H2O2) level and decrease (P < 0.01) in GSH level (vs. control). ET-1 administration slightly downregulated gene expression of p65 of NF- κ B but potently and in a dose-dependent way downregulated p21-cip gene expression in the liver. BAY 11-7082 significantly decreased TBARS (P < 0.001), H2O2 (P < 0.01) and improved the redox status (P < 0.05), compared to ET-1 group. The concentration of TNF- α was increased in the presence of ET-1 (P < 0.05), while BAY 11-7082 decreased TNF- α concentration (P < 0.01). Inhibition of IkB α before ET-1 administration downregulated gene expression of p21-cip but had no effect on p65.
Related Materials