EMAIL THIS PAGE TO A FRIEND
JAMA ophthalmology
Fundus autofluorescence in hydroxychloroquine toxicity.
PMID 23579602
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0915
Hydroxychloroquine sulfate, ≥98% (HPLC), powder
C18H26ClN3O · H2SO4