EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
The effect of amniotic membrane de-epithelialization method on its biological properties and ability to promote limbal epithelial cell culture.
PMID 23580491
Abstract
We characterized the de-epithelialized human amniotic membrane (HAM), and compared cell attachment and proliferation efficiencies. HAM was de-epithelialized by 20% ethanol (AHAM), 1.2 U/mL Dispase (DHAM), 0.02% EDTA (EHAM), 0.25% trypsin-EDTA (THAM), and 5 M urea (UHAM), respectively, followed by gentle scraping with a #15 blade. Surface topology, extracellular matrix (ECM), and growth factor content were characterized and compared to intact HAM by electron microscopies (EM), atomic force microscopy (AFM), immunohistochemistry, and Western blotting. Primary human limbal epithelial cells (LEC) attachment and proliferation efficiencies were assayed. Statistical significance was calculated by SPSS and Fisher's least significant difference test. EHAM, THAM, and UHAM had intact basal lamina and smooth basement membrane surface shown under transmission and scanning EM, and AFM. Cell remnants stayed on AHAM. Disrupted basement membrane and stroma was found in DHAM. Immunostaining intensity quantification and hierarchical clustering revealed that ECM composition of EHAM and UHAM resembled intact HAM. In contrast, DHAM and THAM had drastic loss of ECM and growth factor content. LEC attachment efficiency at 24 hours after seeding was the highest in EHAM (51% as on conventional culture surface), followed by UHAM and AHAM. However, cell proliferation indices at day 10 of culture were similar among different HAM substrates, suggesting repair of ECM and basement membrane by growing epithelial cells. Urea denudation preserved the basement membrane integrity, ECM, and growth factor composition, and had higher cell attachment and proliferation efficiencies. With its short processing time, urea treatment offers a novel alternative for HAM de-epithelialization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ED2SC
Ethylenediaminetetraacetic acid calcium disodium salt
C10H12CaN2Na2O8
E5134
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, for molecular biology, 99.0-101.0% (titration)
C10H14N2Na2O8 · 2H2O
E1644
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E6635
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, plant cell culture tested, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E4884
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
ED2SS
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
C10H14N2Na2O8 · 2H2O
03677
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
C10H14N2Na2O8 · 2H2O
27285
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
PHR1068
Ethylenediaminetetraacetic acid disodium salt dihydrate
C10H14N2Na2O8 · 2H2O
03685
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
C10H14N2Na2O8 · 2H2O
2854
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
C10H14N2Na2O8
E7889
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
C10H14N2Na2O8
T6763 Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
T8003
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
T1005
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
T8658
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
T7309
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
T9935
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
T8802
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
T9201
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
T6424 Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
T0303 Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
T4799 Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
T7409 Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
T5266 Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
T7168 Trypsin from porcine pancreas, tablet, 1 mg tablet
93614 Trypsin from porcine pancreas, ≥10000 U/mg
93615 Trypsin from porcine pancreas, ~1500 U/mg
85450C Trypsin Powder, Porcine 1:250, Gamma irradiated by SER-TAIN Process, dry powder
T3568 TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)