EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Latency-associated degradation of the MRP1 drug transporter during latent human cytomegalovirus infection.
PMID 23580527
Abstract
The reactivation of latent human cytomegalovirus (HCMV) infection after transplantation is associated with high morbidity and mortality. In vivo, myeloid cells and their progenitors are an important site of HCMV latency, whose establishment and/or maintenance require expression of the viral transcript UL138. Using stable isotope labeling by amino acids in cell culture-based mass spectrometry, we found a dramatic UL138-mediated loss of cell surface multidrug resistance-associated protein-1 (MRP1) and the reduction of substrate export by this transporter. Latency-associated loss of MRP1 and accumulation of the cytotoxic drug vincristine, an MRP1 substrate, depleted virus from naturally latent CD14(+) and CD34(+) progenitors, all of which are in vivo sites of latency. The UL138-mediated loss of MRP1 provides a marker for detecting latent HCMV infection and a therapeutic target for eliminating latently infected cells before transplantation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V0400000
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
C46H56N4O10 · H2SO4