EMAIL THIS PAGE TO A FRIEND
Midwifery today with international midwife
Preventing postpartum haemorrhage.
PMID 23581193
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O6379
Oxytocin acetate salt hydrate, ≥97% (HPLC)
C43H66N12O12S2