EMAIL THIS PAGE TO A FRIEND
Tuberkuloz ve toraks
Acute isoniazid intoxication: an uncommon cause of convulsion, coma and acidosis.
PMID 23581267
Abstract
Despite the widespread use, suicidal ingestion of isoniazid is a rare condition in Turkey. We reported a case of acute isoniazid intoxication associated with alcohol intake presenting with convulsion, coma and metabolic acidosis. The patient was treated successfully with intravenous pyridoxine administration. Early recognation and appropriate treatment in the intensive care unit is very important to prevent mortality in patients with acute isoniazid toxicity.