EMAIL THIS PAGE TO A FRIEND
Journal of the American Geriatrics Society
High cortisol and low folate are the only routine blood tests predicting probable Alzheimer's disease after age 75-results of the Vienna Transdanube Aging Study.
PMID 23581921