Description
Molecular Formula
Add to Cart
E0287 Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
E0162 Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
C4749 Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
46051 Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
79302 Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥8.0 U/mg
96667 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
51963 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥0.8 U/mg
46069 Esterase from horse liver, lyophilized, powder, brown, 0.5-1.0 U/mg
46058 Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
E0887 Esterase from rabbit liver, lyophilized powder, ≥75 units/mg protein
79208 Esterase from Rhizopus oryzae, powder, light beige, ≥20 U/g
46071 Esterase from Saccharomyces cerevisiae, lyophilized, powder, beige, ~2 U/g
43388 Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
93855 Esterase Isoenzyme 2 porcine liver, recombinant, recombinant, expressed in E. coli, ≥80 U/g
93712 Esterase Isoenzyme 3 porcine liver, recombinant, recombinant, expressed in E. coli, ≥0.8 U/mg
68998 Esterase Isoenzyme 4 porcine liver, recombinant, recombinant, expressed in E. coli, ≥0.7 U/mg
53564 Esterase Isoenzyme 5 porcine liver, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
80712 Esterase Methylobacterium populi, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
92385 Esterase Nocardia farcinica, recombinant, recombinant, expressed in E. coli, ≥0.4 U/mg
04492 Esterase Paenibacillus barcinonensis, recombinant, recombinant, expressed in E. coli, ≥10 U/mg
04533 Esterase Pelobacter propionicus, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
75742 Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
80711 Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥0.3 U/mg
L0777 Lipase from Aspergillus oryzae, solution, ≥100,000 U/g
62305 Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
62350 Lipozyme®, immobilized from Mucor miehei, granular, brown, >30 U/g