EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
A comprehensive machine-readable view of the mammalian cholesterol biosynthesis pathway.
PMID 23583456
Abstract
Cholesterol biosynthesis serves as a central metabolic hub for numerous biological processes in health and disease. A detailed, integrative single-view description of how the cholesterol pathway is structured and how it interacts with other pathway systems is lacking in the existing literature. Here we provide a systematic review of the existing literature and present a detailed pathway diagram that describes the cholesterol biosynthesis pathway (the mevalonate, the Kandutch-Russell and the Bloch pathway) and shunt pathway that leads to 24(S),25-epoxycholesterol synthesis. The diagram has been produced using the Systems Biology Graphical Notation (SBGN) and is available in the SBGN-ML format, a human readable and machine semantically parsable open community file format.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90469
(RS)-Mevalonic acid lithium salt, ≥96.0% (GC)
C6H11O4 · xLi+
42147
(RS)-Mevalonic acid lithium salt, analytical standard
C6H11O4 · xLi+