EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Chemical modification of erythromycins. IV. Synthesis and biological properties of 6-O-methylerythromycin B.
PMID 2358406
Abstract
6-O-Methylerythromycin B has been synthesized from erythromycin B via regioselective methylation of the 6-hydroxyl group in 71% overall yield. This compound shows in vitro antibacterial activity comparable to erythromycins A and B and exhibits superior in vivo activity with improved pharmacokinetic properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1310000 Erythromycin B, European Pharmacopoeia (EP) Reference Standard
C37H67NO12