EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Direct synthesis of amides from carboxylic acids and amines using B(OCH2CF3)3.
PMID 23586467
Abstract
B(OCH2CF3)3, prepared from readily available B2O3 and 2,2,2-trifluoroethanol, is as an effective reagent for the direct amidation of a variety of carboxylic acids with a broad range of amines. In most cases, the amide products can be purified by a simple filtration procedure using commercially available resins, with no need for aqueous workup or chromatography. The amidation of N-protected amino acids with both primary and secondary amines proceeds effectively, with very low levels of racemization. B(OCH2CF3)3 can also be used for the formylation of a range of amines in good to excellent yield, via transamidation of dimethylformamide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
790877
Tris(2,2,2-trifluoroethyl) borate, 97%
C6H6BF9O3