EMAIL THIS PAGE TO A FRIEND
Current opinion in plant biology
Rice breaks ground for cadmium-free cereals.
PMID 23587938
Abstract
It is important to reduce the risk of long-term cadmium (Cd) exposure through ingestion. Cd in plant-derived foods (especially rice grain in Japan) is a major source of human Cd intake. In rice, Cd uptake and xylem loading in roots, remobilization from leaf blades and intervascular transfer in nodes to redirect Cd transport are crucial for grain Cd accumulation. Different metal transporters mediate these processes with cell-type specific expressions. In this review, we summarize and discuss the recent rapid advance in molecular understanding of Cd transport in rice. The achievements and perspectives in developing 'low-Cd rice' by the use of various alleles of Cd transporter genes are also discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
414891
Cadmium, granular, 30-80 mesh, ≥99%
Cd
265330
Cadmium, granular, ≥99%, 5-20 mesh
Cd
265365
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cd
202886
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cd
348600
Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis
Cd
265454
Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis
Cd
GF61936814
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF33512688
Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF64286602
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF38539929
Cadmium, foil, not light tested, 100x100mm, thickness 0.025mm, 99.7+%
Cd
GF31881911
Cadmium, rod, 500mm, diameter 5.0mm, as drawn, 99.99+%
Cd
GF72757503
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF72955772
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cd
GF85239096
Cadmium, rod, 1000mm, diameter 5.0mm, as drawn, 99.99+%
Cd
GF30096345
Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF59435062
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF01198762
Cadmium, wire reel, 1m, diameter 0.25mm, hard, 99.99+%
Cd
GF82927064
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.999%
Cd
GF32202582
Cadmium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.7+%
Cd
GF86338154
Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cd
GF03378652
Cadmium, rod, 100mm, diameter 5.0mm, as drawn, 99.99+%
Cd
GF50990597
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF12224078
Cadmium, wire reel, 0.2m, diameter 0.25mm, hard, 99.99+%
Cd
GF79039648
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cd
GF37213893
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Cd
05-0075
Cadmium, ≥99.9%
Cd
GF10741889
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF37649100
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF38583287
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF66037754
Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Cd