EMAIL THIS PAGE TO A FRIEND
Circulation
A clinical approach to early repolarization.
PMID 23588960
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5627
Isoprenaline hydrochloride
C11H17NO3 · HCl