EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Enzymatic transformation of nonfood biomass to starch.
PMID 23589840
Abstract
The global demand for food could double in another 40 y owing to growth in the population and food consumption per capita. To meet the world's future food and sustainability needs for biofuels and renewable materials, the production of starch-rich cereals and cellulose-rich bioenergy plants must grow substantially while minimizing agriculture's environmental footprint and conserving biodiversity. Here we demonstrate one-pot enzymatic conversion of pretreated biomass to starch through a nonnatural synthetic enzymatic pathway composed of endoglucanase, cellobiohydrolyase, cellobiose phosphorylase, and alpha-glucan phosphorylase originating from bacterial, fungal, and plant sources. A special polypeptide cap in potato alpha-glucan phosphorylase was essential to push a partially hydrolyzed intermediate of cellulose forward to the synthesis of amylose. Up to 30% of the anhydroglucose units in cellulose were converted to starch; the remaining cellulose was hydrolyzed to glucose suitable for ethanol production by yeast in the same bioreactor. Next-generation biorefineries based on simultaneous enzymatic biotransformation and microbial fermentation could address the food, biofuels, and environment trilemma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6429
α-Cellulose, BioReagent, suitable for insect cell culture
A0512
Amylose from potato, used as amylase substrate
11363
Avicel® PH-101, tested according to Ph.Eur.
22182
Cellulose, acid washed, from spruce, for column chromatography
22184
Cellulose, acid washed, powder, for column chromatography
22183
Cellulose, powder, for column chromatography
22197
Cellulose, DS-0, powder, for thin layer chromatography
09906
Cellulose, DFS-0, microcrystalline, for thin layer chromatography
435244
Cellulose, colloidal, microcrystalline
310697
Cellulose, microcrystalline, powder, 20 μm
435236
Cellulose, microcrystalline, powder
D1523
N,N′-Diacetylchitobiose, ≥96% (HPLC)
C16H28N2O11
85642
Starch, from potato, acc. to Zulkowsky (starch, treated with glycerol at 190°C)
33615
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
03967
Starch, from potato, tested according to Ph.Eur.
S4126
Starch from corn
S9679
Starch from corn, Unmodified waxy corn starch of essentially pure amylopectin; contains only trace amounts of amylose.
S4180
Starch from corn, practical grade
S2388
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
S5296
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
S5651
Starch from potato, for electrophoresis
S2004
Starch from potato, Soluble
S4251
Starch from potato, Powder
S7260
Starch from rice
S5127
Starch from wheat, Unmodified
C6413
Cellulose, microgranular