EMAIL THIS PAGE TO A FRIEND
Anthropologischer Anzeiger; Bericht uber die biologisch-anthropologische Literatur
Dopamine D2 receptor and ankyrin repeat and kinase domain containing 1 genes among the two linguistically distinct tribal populations of Ranchi district, Jharkhand, India.
PMID 23590110
Abstract
In the recent years, haplotype studies have emerged as a critical tool for studying the human migratory patterns. Dopamine D2 receptor (DRD2) and Ankyrin Repeat and Kinase Domain Containing 1 (ANKK1) genes, which also bear specific clinical implications in various neuropsychiatric and behavioural/addictive disorders, are significant nuclear DNA markers for studying human genome diversity. The present study was conducted in order to understand the distribution pattern of the three DRD2 and ANKK1 TaqI sites and also the frequencies of their haplotypes among Oraons (n = 48) and Mundas (n = 50)--the two linguistically distinct tribal population groups of Jharkhand. The phylogenetic inference was drawn through the statistical comparisons of the present DRD2 and ANKK1 TaqI site data with the available data from population groups belonging to other parts of India and also rest of the world (ALFRED Database). All the three TaqI sites were found to be polymorphic among Oraons and Mundas with relatively high average heterozygosities. Oraons exhibited a comparatively higher frequency of the ancestral B2D2A1 haplotype (0.356) than the Mundas (0.193). Significant and higher linkage disequilibrium (LD) values between all three sites were observed among Mundas which is indicative of admixture, whereas Oraons exhibited non significant and low LD values. The presence of ancestral haplotype B2D2A1 in higher frequency and lower and non-significant LD among Oraons suggest that they might be the older inhabitants in the region though the major limitation of the study is small sample size which might have introduced bias in the accuracy of the calculated pairwise LD for the three polymorphic sites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R6142 Acc I from Acinetobacter calcoaceticus, Restriction Enzyme
R6885 Alu I from Arthrobacter luteus, Restriction Enzyme
R8631
Bcl I from Bacillus caldolyticus, Restriction Enzyme
R6377
Bgl II from Bacillus licheniformis, Restriction Enzyme
R3131
Bln I from Brevibacterium linens, buffered aqueous glycerol solution
R3635
Bsm I from Bacillus stearothermophilus NUB 36, Restriction Enzyme
R4253 BstE II from Bacillus stearothermophilus ET, Restriction Enzyme
R1761
Cfo I from Clostridium formicoaceticum, Restriction Enzyme
R7763
Cla I from Caryophanon latum L, Restriction Enzyme
R4256
Dde I from Desulfovibrio desulfuricans strain Norway, Restriction Enzyme
R4381
Dra I from Deinococcus radiophilus, Restriction Enzyme
R3884 EclX I from Enterobacter cloacae 590, Restriction Enzyme
R6265
EcoR I from Escherichia coli BS5, Restriction Enzyme
R2756
EcoR V from Escherichia coli, buffered aqueous glycerol solution
R5628 Hae III from Haemophilus aegyptius, Restriction Enzyme
R1632
Mva I from Micrococcus varians Rfl 19, Restriction Enzyme
R8761 Nco I from Nocardia corallina, Restriction Enzyme
R5634
Nhe I from Neisseria mucosa heidelbergensis, Restriction Enzyme
R8506
Not I from Nocardia otidiscaviarum, Restriction Enzyme
R5884 Nsi I from Neisseria sicca, Restriction Enzyme
R7023
Pst I from Providencia stuartii, Restriction Enzyme
R1508
Pvu I from Proteus vulgaris, Restriction Enzyme
R2631
Pvu II from Proteus vulgaris, Restriction Enzyme
R4756 Rsa I from Rhodopseudomonas sphaeroides, Restriction Enzyme
R0754 Sal I from Streptomyces albus G, Restriction Enzyme
R8256
Sfi I from Streptomyces fimbriatus, Restriction Enzyme
R7135 Sph I from Streptomyces phaeochromogenes, Restriction Enzyme
R7260
Xba I from Xanthomonas badrii, Restriction Enzyme
R6379
Xho I from Xanthomonas holcicola, Restriction Enzyme
R1259 Afl III from Anabaena flos-aquae, buffered aqueous glycerol solution