EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
β-Secretase (BACE1) inhibitors with high in vivo efficacy suitable for clinical evaluation in Alzheimer's disease.
PMID 23590342
Abstract
An extensive fluorine scan of 1,3-oxazines revealed the power of fluorine(s) to lower the pKa and thereby dramatically change the pharmacological profile of this class of BACE1 inhibitors. The CF3 substituted oxazine 89, a potent and highly brain penetrant BACE1 inhibitor, was able to reduce significantly CSF Aβ40 and 42 in rats at oral doses as low as 1 mg/kg. The effect was long lasting, showing a significant reduction of Aβ40 and 42 even after 24 h. In contrast to 89, compound 1b lacking the CF3 group was virtually inactive in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4562 β-Secretase inhibitor, ≥97% (HPLC), powder
C73H118N16O27