EMAIL THIS PAGE TO A FRIEND
Heart (British Cardiac Society)
Periprocedural type IVa myocardial infarction and the importance of platelet inhibition.
PMID 23591669
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3013
Adenosine 2′(3′)-monophosphate mixed isomers
C10H14N5O7P