EMAIL THIS PAGE TO A FRIEND
Journal of molecular medicine (Berlin, Germany)
Rehabilitating rimonabant.
PMID 23591956
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0800 Rimonabant hydrochloride, ≥98% (HPLC)
C22H21Cl3N4O · HCl