EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Endocrinology and Metabolism
Upregulation of p21 activates the intrinsic apoptotic pathway in ?-cells.
PMID 23592481
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-536
Anti-Bak Antibody, NT, clone, Upstate®, from rabbit