EMAIL THIS PAGE TO A FRIEND
The journal of adhesive dentistry
Influence of peripheral enamel bonding and chlorhexidine pretreatment on resin bonding to dentin.
PMID 23593635
Abstract
To evaluate the influence of peripheral enamel bonding, chlorhexidine digluconate (CHX) pretreatment, and storage time on resin-dentin microtensile bond strength (µTBS) of a self-etching adhesive system and selfetching cement. Fifty-six noncarious human third molars were coronally sectioned to provide a flat dentin surface; the circumferential enamel was removed from half of these and the other half was left intact. Pretreatment with 0.2% or 2% CHX or none (control) was performed before bonding composite blocks with Panavia F2.0 or RelyX Unicem to dentin. Specimens were stored in 0.5% chloramine for 3 days (control) or 6 months. Bonded specimens were sectioned into 1-mm2 dentin-only beams and a tensile load was applied until failure. The resulting 14 test groups, defined by control group/storage time/cement/CHX application mode/enamel bonding, were analyzed using two-way and three-way ANOVA, as well as Tukey's HSD post-hoc test (α = 0.05). Failure patterns of the specimens were observed and measured using scanning electron microscopy. The comparison of the control vs experimental subgroups with peripheral enamel bonding and without CHX pretreatment after six months found no significant differences regarding the µTBS between the cements (p > 0.05) but did for the storage time (p <= 0.05). When storing the specimens for six months, the absence of peripheral enamel bonding significantly decreased the µTBS (p <= 0.05). Panavia F2.0 specimens showed significantly higher µTBS values than RelyX Unicem specimens (p <= 0.05). CHX pretreatment, regardless the concentration, showed a significant influence on the µTBS (p <= 0.05). SEM revealed that peripheral enamel bonding reduced failures at the resin/dentin interface. The absence of peripheral enamel bonding and a longer storage time decrease the µTBS of twostep self-etching adhesive system and a one-step self-etching cement. CHX pretreatment of the dentinal surfaces seems to improve the durability of the bond to dentin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1540000
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
C22H30Cl2N10 · 2HCl