EMAIL THIS PAGE TO A FRIEND
The Journal of bone and joint surgery. American volume
Immunohistochemical demonstration of cyclooxygenase-2 in glomus tumors.
PMID 23595071
Abstract
Glomus tumors are benign hamartomas that account for 1% to 5% of all soft-tissue tumors of the hand. Painful spasms radiating from the lesion are typical clinical signs. As the pain production mechanism is unclear, we evaluated S100 protein, substance P, and cyclooxygenase-2 expression by immunohistochemistry in glomus tumor samples. Eight solitary glomus tumors were surgically excised and confirmed histologically by an experienced pathologist. Paraffin-embedded tissues were prepared for immunohistochemistry. The sections were stained with separate polyclonal antibodies for S100, substance P, and cyclooxygenase-2. In three of the tumors, we measured the prostaglandin E2 concentrations. All samples were positive for S100 protein and cyclooxygenase-2.Substance P was found in five of the eight samples. High prostaglandin-E2 concentrations were seen in all three samples tested. Cyclooxygenase-2-immunoreactive cells are present in solitary glomus tumors. Since cyclooxygenase-2 produces prostaglandin E2, which is thought to be a strong vasodilator, the pain could be caused by vasodilation in the glomus tumor, with increased intracapsular pressure.