EMAIL THIS PAGE TO A FRIEND
The Journal of nutrition
Sociodemographic and lifestyle variables are compound- and class-specific correlates of urine phytoestrogen concentrations in the U.S. population.
PMID 23596167
Abstract
Isoflavones and lignans are plant-derived dietary compounds generally believed to be beneficial to human health. We investigated the extent to which sociodemographic (age, sex, race-ethnicity, education, and income) and lifestyle variables (smoking, alcohol consumption, BMI, physical activity, and dietary supplement use) were correlates of spot urine concentration for daidzein, genistein, O-desmethylangolensin (DMA), equol, enterodiol, and enterolactone in the U.S. population aged ≥ 20 y (NHANES 2003-2006). We performed correlation analyses with continuous variables and calculated stratified unadjusted geometric means for each sociodemographic and lifestyle variable. We used bivariate significance testing and covariate adjustment by use of multiple regression models to identify influential variables and used β coefficients to estimate relative effects. Urine creatinine was also included in our analyses because of its use in correcting for variable dilution in spot urine samples. We observed many significant (P < 0.05) associations with the sociodemographic and lifestyle variables that withstood covariate adjustment. Smoking was a significant correlate of urine DMA and enterolactone, with concentrations at least 25% lower in smokers vs. nonsmokers. Consumers of 1 daily alcoholic drink vs. none were estimated to have 18-21% lower urine equol and DMA concentrations. A 25% increase in BMI was associated with a 21% lower urine enterolactone concentration, and increasing physical activity was associated with a >6% higher urine enterolactone concentration. Dietary supplement use was not significantly associated with any of the urine phytoestrogens. Overall, we found that relationships between sociodemographic and lifestyle variables and urine phytoestrogen concentration were highly compound and class specific.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45199
Enterolactone, ~95% (HPLC)
C18H18O4