EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical constituents from Daphne acutiloba].
PMID 23596879
Abstract
To study the chemical constituents from Daphne acutiloba. The constituents were separated by column chromatography, and their structures were elucidated by spectroscopic data analyses. Fifteen compounds were isolated from the EtOAc extract and identified as wikstroelide M (1), vesiculosin (2), prostratin (3), 7-hydroxy-coumarin (4), 7,8-di-hydroxy-coumarin (5), isodaphnoside (6), daphnorine (7), rutamontine (8), daphnolin (9), daphneticin (10), (+)-pinoresinol-beta-D-glucoside (11), oleodapnone (12), oleodaphnal (13), ergosterol peroxide (14) and cholest-5-en-3beta-ol (15). All the compounds except for 4, 5 and 14 were obtained from the stems of this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40674
Pinoresinol, ≥95.0% (HPLC)
C20H22O6
06709
Pinoresinol, analytical standard
C20H22O6