EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
A conversation with Jeffrey M. Friedman by Ushma S. Neill.
PMID 23598296
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4429 Pancreozymin from porcine intestine, 2-6 Crick units/mg solid