EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis of new 1,2,3-triazole derivatives of uracil and thymine with potential inhibitory activity against acidic corrosion of steels.
PMID 23599018
Abstract
Ten 1,4-disubstituted 1,2,3-triazoles were synthesized from one of 1-(azido-methyl)benzene, 1-(azidomethyl)-4-fluorobenzene, 1-(azidomethyl)-4-chlorobenzene, 1-(azidomethyl)-4-bromobenzene or 1-(azidomethyl)-4-iodobenzene, generated in situ from sodium azide and the corresponding benzyl halide, and dipropargyl uracil or dipropargyl thymine. Optimal experimental conditions were established for the conventional click chemistry. The corrosion inhibiting properties of some of these compounds, which were determined by means of an electrochemical technique, are also presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
515604
4-Iodobenzyl bromide, 95%
C7H6BrI