EMAIL THIS PAGE TO A FRIEND
British journal of anaesthesia
Fade of train-of-four ratio despite administration of more than 12 mg kg(-1) sugammadex in a myasthenia gravis patient receiving rocuronium.
PMID 23599531
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000527
Rocuronium for peak identification, European Pharmacopoeia (EP) Reference Standard
C32H53BrN2O4