EMAIL THIS PAGE TO A FRIEND
Tropical gastroenterology : official journal of the Digestive Diseases Foundation
'Myiasis-in-ano'.
PMID 23600060
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W308900 L-Turpentine, FG
24245 Oil of turpentine, purified