EMAIL THIS PAGE TO A FRIEND
Kinderkrankenschwester : Organ der Sektion Kinderkrankenpflege
[Study indites calcium as a heart breaker. 30 % more myocardial infarcts caused by high dosage calcium preparations].
PMID 23600116