EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Two new glycosides from the florets of Carthamus tinctorius.
PMID 23600648
Abstract
Two new glycoside compounds, named saffloquinoside C (1) and (-)-4-hydroxybenzoic acid-4-O-[6'-O-(2″-methylbutyryl)-β-D-glucopyranoside] (2), were isolated from the florets of Carthamus tinctorius. Their structures were elucidated by detailed spectroscopic means including UV, IR, HR-ESI-MS, 1D and 2D NMR, and CD data. Compound 1 was a rare quinochalcone glycoside with six five-membered dioxaspirocycle.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3766
Sodium 4-hydroxybenzoate, analytical standard
C7H5NaO3