EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Two new tigliane diterpene esters from the flower buds of Daphne genkwa.
PMID 23600859
Abstract
The phytochemical investigation of the flower buds of Daphne genkwa yielded four highly oxygenated tigliane diterpene esters (1-4), including two new phorbol derivatives, 12-O-(2'E,4'E-decadienoyl)-7-oxo-5-ene-phorbol-13-acetate (1) and 12-O-neodecanoyl-7-oxo-5-ene-phorbol-13-acetate (2). The molecular structures of the isolated compounds were elucidated on the basis of extensive spectroscopic analysis, including 1D and 2D NMR.