EMAIL THIS PAGE TO A FRIEND
Life sciences
Nefiracetam attenuates post-ischemic nonconvulsive seizures in rats and protects neuronal cell death induced by veratridine and glutamate.
PMID 23603142
Abstract
Stroke patients are at a high risk of developing post-ischemic seizures and cognitive impairment. Nefiracetam (NEF), a pyrrolidone derivative, has been shown to possess both anti-epileptic and cognitive-enhancing properties. In this study the anti-seizure effects of NEF were evaluated in a rat model of post-ischemic nonconvulsive seizures (NCSs). Its potential mechanisms were investigated in neuronal cell culture assays of neurotoxicity associated with ischemic brain injury and epileptogenesis. In the in vivo study, rats received 24h permanent middle cerebral artery occlusion. NEF was administered intravenously either at 15 min post-injury but prior to the first NCS event (30 mg/kg, pre-NCS treatment) or immediately after the first NCS occurred (30 or 60 mg/kg, post-NCS treatment). In the in vitro study, neuronal cell cultures were exposed to veratridine or glutamate and treated with NEF (1-500 nM). The NEF pre-NCS treatment significantly reduced the NCS frequency and duration, whereas the higher NEF dose (60 mg/kg) was required to achieve similar effects when given after NCS occurred. The NEF treatment also dose-dependently (5-500 nM) protected against neuronal cell death induced by veratridine as measured by MTT cell viability assay, but higher doses (250-500 nM) were required against glutamate toxicity. The anti-seizure property of NEF was demonstrated in a clinically relevant rat model of post-ischemic NCS. The preferential effects of NEF against in vitro veratridine toxicity suggest the involvement of its modulation of sodium channel malfunction. Future studies are warranted to study the mechanisms of NEF against ischemic brain injury and post-ischemic seizures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N2288
Nefiracetam, solid
C14H18N2O2