EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Effects of endocrine disrupting chemicals on in vitro global DNA methylation and adipocyte differentiation.
PMID 23603478
Abstract
Recent studies suggest that endocrine disrupting chemicals (EDCs) may form a risk factor for obesity by altering energy metabolism through epigenetic gene regulation. The goal of this study is to investigate the effects of a range of EDCs with putative obesogenic properties on global DNA methylation and adipocyte differentiation in vitro. Murine N2A and human SK-N-AS neuroblastoma cells and murine preadipocyte fibroblasts (3T3-L1) were exposed to tributyltin (TBT), diethylstilbestrol (DES), bisphenol A (BPA), 2,3,7,8-tetrachlorodibenzo-[p]-dioxin (TCDD), 2,2',4,4',5,5'-hexachlorobiphenyl (PCB-153), hexachlorobenzene (HCB), hexabromocyclododecane (HBCD), 2,2',4,4'-tetrabrominated diphenyl ether (BDE-47) , perfluorinated octyl acid (PFOA) and perfluorinated octyl sulfonate (PFOS). A modest decrease in global DNA methylation was observed in N2A cells exposed to 10 μM DES, BPA, TCDD, BDE-47, PCB-153 and 1 μM HCB, but no changes were found in the human SK-N-AS cells. We reveal for the first time that BDE-47 increases adipocyte differentiation in a dose-dependent manner (2.5-25 μM). Adipocyte differentiation was also enhanced by TBT (≥ 10 nM) and BPA (>10 μM) and inhibited by TCDD (≥ 0.1 nM). The other chemicals showed either modest or no effects on adipocyte differentiation at the concentrations tested (PFOA, PFOS and HBCD at 10 μM; PCB-153, 3.4 μM and HCB, 1 μM). This study demonstrates that selected EDCs can induce functional changes in murine adipocyte differentiation in vitro which are accompanied by decreased global DNA methylation.